Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hoxb13
Entrez Gene ID
100773981
Gene Name
homeobox B13
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613745: 36236 - 38027 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499550.1 XP_003499598.1 1 - 2 601 62

View Full Product Details