Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zc3h12c
Entrez Gene ID
100753033
Gene Name
zinc finger CCCH-type containing 12C
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613912: 244712 - 273935 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643376.1 XP_007641566.1 3 - 4 1358 108
GenBank mRNA JI876926.1 - 3 - 4 566 108

View Full Product Details