Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tppp2
Entrez Gene ID
100758674
Gene Name
tubulin polymerization-promoting protein family member 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614416: 774623 - 776541 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507718.2 XP_003507766.2 2 - 3 326 97

View Full Product Details