Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC100764822
Entrez Gene ID
100764822
Gene Name
NUT family member Gm806-like
Gene Aliases
I79_024614
Chromosome Location
Chr.Un|NW_003617477: 15266 - 22628 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007655170.1 XP_007653360.1 4 - 5 1051 97

View Full Product Details