Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hoxb4
Entrez Gene ID
100750374
Gene Name
homeobox B4
Gene Aliases
I79_005670
Chromosome Location
Chr.Un|NW_003613745: 155508 - 157092 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499554.2 XP_003499602.1 1 - 1 421 128

View Full Product Details