Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PMVK
Entrez Gene ID
102172312
Gene Name
phosphomevalonate kinase
Gene Aliases
-
Chromosome Location
Chr.3: 104466965 - 104478956 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005677425.3 XP_005677482.1 1 - 2 569 54
GenBank mRNA JT645579.1 - 1 - 2 153 54

View Full Product Details