Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DUSP27
Entrez Gene ID
102189918
Gene Name
dual specificity phosphatase 27, atypical
Gene Aliases
-
Chromosome Location
Chr.3: 118441217 - 118476328 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018046440.1 XP_017901929.1 1 - 2 268 58
GenBank mRNA JO348517.2 - 3 - 4 568 58
JO348518.2 - 3 - 4 568 58

View Full Product Details