Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLR3G
Entrez Gene ID
102178067
Gene Name
RNA polymerase III subunit G
Gene Aliases
-
Chromosome Location
Chr.7: 20457562 - 20509102 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005683428.3 XP_005683485.1 4 - 5 458 122
XM_018050046.1 XP_017905535.1 4 - 5 458 122

View Full Product Details