The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005689562.3 | XP_005689619.2 | 2 - 3 | 352 | 117 |
GenBank mRNA | JO634883.2 | - | 2 - 3 | 373 | 117 |