Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARL4C
Entrez Gene ID
102191580
Gene Name
ADP ribosylation factor like GTPase 4C
Gene Aliases
-
Chromosome Location
Chr.3: 6672189 - 6676210 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018040905.1 XP_017896394.1 1 - 1 1049 57
GenBank mRNA JT687701.1 - 7 - 7 2793 57

View Full Product Details