Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH3RF3
Entrez Gene ID
102178914
Gene Name
SH3 domain containing ring finger 3
Gene Aliases
-
Chromosome Location
Chr.11: 43939878 - 44099310 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018055296.1 XP_017910785.1 2 - 3 1770 74
XM_018055297.1 XP_017910786.1 2 - 3 1770 74

View Full Product Details