Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDC5L
Entrez Gene ID
100861007
Gene Name
cell division cycle 5 like
Gene Aliases
-
Chromosome Location
Chr.23: 30940872 - 30985913 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005696360.3 XP_005696417.1 4 - 5 563 63
GenBank mRNA JI861662.1 - 4 - 5 434 63

View Full Product Details