Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
FAM71E2
Entrez Gene ID
102184653
Gene Name
family with sequence similarity 71 member E2
Gene Aliases
-
Chromosome Location
Chr.18: 63882140 - 63888338 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018063236.1 XP_017918725.1 8 - 8 2048 74

View Full Product Details