Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZW10
Entrez Gene ID
102178922
Gene Name
zw10 kinetochore protein
Gene Aliases
-
Chromosome Location
Chr.15: 58041779 - 58076745 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005689464.2 XP_005689521.2 5 - 6 668 70

View Full Product Details