The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_013969002.2 | XP_013824456.1 | 3 - 4 | 1657 | 102 |
XM_018064344.1 | XP_017919833.1 | 3 - 4 | 1776 | 102 | |
XM_018064349.1 | XP_017919838.1 | 3 - 4 | 1400 | 102 | |
GenBank mRNA | JO375717.2 | - | 1 - 2 | 526 | 102 |