Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC3A
Entrez Gene ID
102179125
Gene Name
C-type lectin domain family 3 member A
Gene Aliases
-
Chromosome Location
Chr.18: 6434852 - 6443847 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005691819.3 XP_005691876.3 1 - 2 343 69

View Full Product Details