Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102190786
Entrez Gene ID
102190786
Gene Name
NUT family member 2G
Gene Aliases
NUTM2G
Chromosome Location
Chr.8: 89926735 - 89932878 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005684379.2 XP_005684436.2 2 - 3 779 68

View Full Product Details