Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mtt
Entrez Gene ID
35832
Gene Name
mangetout
Gene Aliases
CG18447, CG30361, CG8692, DmGluBR, DmGluRB, DmXR, Dmel\CG30361, Dmel_CG30361, Glu-RB, GluRB, anon-WO0170980.4, anon-WO0170980.5, mXr
Chromosome Location
Chr.2R: 8473956 - 8532294 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0050361
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144147.2 NP_001137619.1 8 - 9 3736 70
NM_001273877.1 NP_001260806.1 8 - 9 3736 70
NM_136542.3 NP_610386.2 7 - 8 2776 70
NM_165609.3 NP_724683.2 6 - 7 1830 70
NM_206058.2 NP_995780.1 2 - 3 302 70
GenBank mRNA BT033068.1 - 2 - 3 303 70

View Full Product Details