Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tsp29Fb
Entrez Gene ID
34212
Gene Name
Tetraspanin 29Fb
Gene Aliases
CG9496, Dm.Tsp29Fb, Dmel\CG9496, Dmel_CG9496, anon-EST:ParkEST130
Chromosome Location
Chr.2L: 8937390 - 8939743 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032075
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144337.2 NP_001137809.1 1 - 2 248 91
NM_078792.3 NP_523516.1 2 - 3 134 91
NM_164838.2 NP_723426.1 2 - 3 177 91
GenBank mRNA AF274012.1 - 2 - 3 134 91
AY122216.1 - 2 - 3 134 91

View Full Product Details