Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
scaf
Entrez Gene ID
35505
Gene Name
scarface
Gene Aliases
CG11066, CG15900, Dmel\CG11066, Dmel_CG11066, SPH142, Scaf, cSPH142, scarf
Chromosome Location
Chr.2R: 5676750 - 5691723 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033033
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136336.3 NP_610180.1 6 - 7 1786 91
NM_165441.2 NP_724424.1 6 - 7 2133 91
GenBank mRNA AY047548.1 - 6 - 7 1758 91

View Full Product Details