Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
emp
Entrez Gene ID
37999
Gene Name
epithelial membrane protein
Gene Aliases
CG2727, Dmel\CG2727, Dmel_CG2727, Emp
Chromosome Location
Chr.2R: 24976456 - 24986070 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010435
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299943.1 NP_001286872.1 5 - 6 1288 93
NM_001299944.1 NP_001286873.1 5 - 6 1717 93
NM_079135.4 NP_523859.2 5 - 6 1307 93
NM_166702.3 NP_726504.2 5 - 6 1274 93
NM_166703.3 NP_726505.1 6 - 7 1487 93

View Full Product Details