Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- tum
- Entrez Gene ID
-
36538
- Gene Name
- tumbleweed
- Gene Aliases
-
13345, AcGAP, CG13345, DRacGAP, Dmel\CG13345, Dmel_CG13345, RacGAP, RacGAP50C, RacGAP50c, RacGap, RacGap50C, RhoGAP-50C14, TUM, Tum, Tumbleweed, acGAP, eon, i249, racGAP50, racGAP50C, racGAP50c
- Chromosome Location
-
Chr.2R: 13856150 - 13858689 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0086356
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137068.4 |
NP_610912.2 |
1 - 2 |
284 |
62 |
GenBank mRNA |
BT150442.1 |
- |
1 - 2 |
312 |
62 |
View Full Product Details