Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atg9
Entrez Gene ID
36821
Gene Name
Autophagy-related 9
Gene Aliases
ATG9, CG3615, Dmel\CG3615, Dmel_CG3615, atg9, dAtg9
Chromosome Location
Chr.2R: 16294400 - 16297232 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034110
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274094.2 NP_001261023.1 1 - 1 2662 74
NM_137270.4 NP_611114.1 1 - 1 2662 74
GenBank mRNA BT006028.1 - 1 - 1 2660 74

View Full Product Details