Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- tutl
- Entrez Gene ID
-
46015
- Gene Name
- turtle
- Gene Aliases
-
CG15426, CG15427, CG15428, CT35489, CT35490, Dmel\CG15427, Dmel_CG15427, Tutl, jf4, l(2)01085, l(2)24Df, l(2)jf4, mdcds_7441
- Chromosome Location
-
Chr.2L: 4283148 - 4323781 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0010473
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080127.6 |
NP_524866.4 |
10 - 11 |
3162 |
76 |
View Full Product Details