Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fie
Entrez Gene ID
38452
Gene Name
Fire exit
Gene Aliases
BEST:HL04053, CG1079, Dmel\CG1079, Dmel_CG1079, HL04053
Chromosome Location
Chr.3L: 3892574 - 3894833 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026592
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274452.1 NP_001261381.1 3 - 4 780 64
NM_133165.2 NP_573393.1 3 - 4 937 64
GenBank mRNA AY060992.1 - 3 - 4 758 64

View Full Product Details