Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
grsm
Entrez Gene ID
41587
Gene Name
granny smith
Gene Aliases
BcDNA:LD41548, CG7340, Dmel\CG7340, Dmel_CG7340, granny-smith, l(3)c00064
Chromosome Location
Chr.3R: 12950042 - 12967712 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040493
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141983.3 NP_650240.1 1 - 2 208 73
GenBank mRNA AF218811.1 - 1 - 2 177 73
BT100045.1 - 1 - 2 140 73

View Full Product Details