Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bili
Entrez Gene ID
43019
Gene Name
Band4.1 inhibitor LRP interactor
Gene Aliases
CG11848, Dmel\CG11848, Dmel_CG11848, dBili
Chromosome Location
Chr.3R: 25131306 - 25136709 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039282
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170251.2 NP_001163722.1 8 - 9 2200 59
NM_001170252.2 NP_001163723.1 8 - 9 2189 59
NM_001170253.2 NP_001163724.1 8 - 9 2284 59
NM_001170254.2 NP_001163725.1 8 - 9 2220 59
NM_001170255.1 NP_001163726.1 8 - 9 2340 59
NM_001300592.1 NP_001287521.1 8 - 9 2593 59
NM_143085.3 NP_651342.3 8 - 9 2297 59
GenBank mRNA AY094737.1 - 8 - 9 2259 59

View Full Product Details