Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- grsm
- Entrez Gene ID
-
41587
- Gene Name
- granny smith
- Gene Aliases
-
BcDNA:LD41548, CG7340, Dmel\CG7340, Dmel_CG7340, granny-smith, l(3)c00064
- Chromosome Location
-
Chr.3R: 12950042 - 12967712 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0040493
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_169482.1 |
NP_731749.3 |
1 - 2 |
263 |
65 |
- Gene Symbol
- CG14384
- Entrez Gene ID
-
41588
- Gene Name
- CG14384-PA
- Gene Aliases
-
Dmel\CG14384, Dmel_CG14384
- Chromosome Location
-
Chr.3R: 12965938 - 12967712 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038097
View Full Product Details