Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nAChRbeta2
Entrez Gene ID
42920
Gene Name
nicotinic Acetylcholine Receptor beta2
Gene Aliases
Acr96Ac, AcrF, CG6798, Dbeta2, Dmel\CG6798, Dmel_CG6798, EMS2, SBD, beta2, beta[[2]], nACR beta 96A, nACRbeta, nAChR, nAChR-beta96A, nAcRbeta, nAcRbeta-96A, nAcRs, nAcr96Ac, sbd
Chromosome Location
Chr.3R: 24509228 - 24515106 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004118
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079759.4 NP_524483.1 5 - 6 829 86
NM_170147.3 NP_733002.1 5 - 6 829 86
GenBank mRNA BT133158.1 - 5 - 6 809 86

View Full Product Details