Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ETHR
- Entrez Gene ID
-
42523
- Gene Name
- CG5911-PA
- Gene Aliases
-
CG5911, CG5911a, CG5911b, Dm ETHRa, Dmel\CG5911, Dmel_CG5911, anon-WO0170980.37, anon-WO0170980.38
- Chromosome Location
-
Chr.3R: 21247011 - 21265134 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038874
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_206533.2 |
NP_996255.1 |
4 - 5 |
1603 |
60 |
GenBank mRNA |
AY220742.1 |
- |
3 - 4 |
436 |
60 |
View Full Product Details