Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CrebB
Entrez Gene ID
32817
Gene Name
Cyclic-AMP response element binding protein B
Gene Aliases
CG6103, CRE-BP, CREB, CREB-B, CREB-b, CREB2, CREB2-b, CREB2a, CREB2b, Creb, Creb B 17A, Creb17A, Creb2, Creb2-b, Creb2.b, Creb2b, CrebB-17A, CrebB-17B, CrebB17-A, DM9, Dmel\CG6103, Dmel_CG6103, S162, dCREB, dCREB-2, dCREB-2a, dCREB-B, dCREB2, dCREB2-a, dCREB2-b, dCREB2a, dCREB2b, dCbz, dCreb2, dCreb2-b, dCreb2b, dCrebB2, dcreb2-b, l(1)17Af
Chromosome Location
Chr.X: 18366357 - 18377864 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0265784
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103546.3 NP_001097016.2 4 - 5 948 89
NM_001103547.4 NP_001097017.1 3 - 4 1190 89
NM_001103548.4 NP_001097018.1 3 - 4 1190 89
NM_001272748.3 NP_001259677.1 3 - 4 770 89
NM_001272749.2 NP_001259678.1 4 - 5 906 89
NM_001382186.1 NP_001368947.1 3 - 4 1194 89
NM_206783.4 NP_996506.2 3 - 4 917 89

View Full Product Details