Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
phactr3b
Entrez Gene ID
553558
Gene Name
phosphatase and actin regulator 3b
Gene Aliases
zgc:109967
Chromosome Location
Chr.11: 1422737 - 1509775 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020537.1 NP_001018373.1 2 - 3 360 118
XM_009305736.3 XP_009304011.1 2 - 3 362 118
XM_021479616.1 XP_021335291.1 2 - 3 362 118
XM_021479617.1 XP_021335292.1 2 - 3 361 118
GenBank mRNA BC095090.1 - 2 - 3 360 118

View Full Product Details