Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
valopa
Entrez Gene ID
58109
Gene Name
vertebrate ancient long opsin a
Gene Aliases
(VA)-opsin, VA-opsin, VAL-opsin, va1, valop
Chromosome Location
Chr.13: 51502947 - 51527140 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131586.2 NP_571661.1 3 - 4 901 82
GenBank mRNA AB035276.1 - 3 - 4 668 82
AB035277.1 - 3 - 4 668 82
BC128815.1 - 3 - 4 901 82
KT008402.1 - 3 - 4 82

View Full Product Details