Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rer1
Entrez Gene ID
393648
Gene Name
retention in endoplasmic reticulum sorting receptor 1
Gene Aliases
zgc:65968
Chromosome Location
Chr.8: 48942479 - 48959611 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200675.2 NP_956969.1 5 - 6 405 60
XM_005167296.4 XP_005167353.2 5 - 6 487 60
XM_005167298.4 XP_005167355.2 5 - 6 472 60
XM_005167299.4 XP_005167356.1 5 - 6 492 60
GenBank mRNA AY398406.1 - 5 - 6 405 60
BC050162.1 - 5 - 6 497 60
BC058292.1 - 5 - 6 407 60

View Full Product Details