Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
prpf6
Entrez Gene ID
323855
Gene Name
PRP6 pre-mRNA processing factor 6 homolog (S. cerevisiae)
Gene Aliases
c20orf14, fc12b02, wu:fa05f07, wu:fc12b02, zgc:65913
Chromosome Location
Chr.23: 7000173 - 7052386 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212655.2 NP_997820.2 9 - 10 1339 82
GenBank mRNA BC056710.1 - 9 - 10 1315 82
BC066556.1 - 9 - 10 1303 82

View Full Product Details