Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
irf2a
Entrez Gene ID
325410
Gene Name
interferon regulatory factor 2a
Gene Aliases
wu:fc74h04, zgc:76951
Chromosome Location
Chr.1: 39865650 - 39880260 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001326712.1 NP_001313641.1 3 - 4 432 78
NM_207054.2 NP_996937.2 3 - 4 432 78
GenBank mRNA BC066487.1 - 3 - 4 329 78

View Full Product Details