Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pycr3
Entrez Gene ID
560682
Gene Name
pyrroline-5-carboxylate reductase 3
Gene Aliases
pycrl, si:dkey-235d18.3
Chromosome Location
Chr.20: 52529093 - 52541512 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044938.1 NP_001038403.1 2 - 3 181 76
GenBank mRNA BC124544.1 - 2 - 3 105 76
BC135053.1 - 2 - 3 230 76

View Full Product Details