Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
desmb
Entrez Gene ID
768287
Gene Name
desmin b
Gene Aliases
wu:fc11d08, zgc:154009
Chromosome Location
Chr.6: 24147 - 44434 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077452.1 NP_001070920.1 1 - 2 623 69
XM_021476935.1 XP_021332610.1 1 - 2 645 69
GenBank mRNA BC124512.1 - 1 - 2 623 69
BC165598.1 - 1 - 2 618 69

View Full Product Details