Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
med10
Entrez Gene ID
550394
Gene Name
mediator complex subunit 10
Gene Aliases
png, zgc:112222
Chromosome Location
Chr.19: 27970362 - 27974152 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017699.1 NP_001017699.1 1 - 2 241 84
GenBank mRNA AY698065.1 - 1 - 2 84
BC093274.1 - 1 - 2 241 84

View Full Product Details