Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
blk
Entrez Gene ID
567764
Gene Name
BLK proto-oncogene, Src family tyrosine kinase
Gene Aliases
bltk, si:dkey-33i22.2, zgc:136231
Chromosome Location
Chr.20: 19176443 - 19196138 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030220.1 NP_001025391.1 12 - 13 1197 115
XM_017352511.2 XP_017208000.1 11 - 12 1797 115

View Full Product Details