Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
col8a2
Entrez Gene ID
794479
Gene Name
collagen, type VIII, alpha 2
Gene Aliases
si:ch211-197n10.4
Chromosome Location
Chr.19: 35496282 - 35596207 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082901.1 NP_001076370.1 2 - 3 550 68

View Full Product Details