Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kirrel3l
Entrez Gene ID
557315
Gene Name
kirre like nephrin family adhesion molecule 3, like
Gene Aliases
cb33, kirrel3, sb:cb33, si:dkey-29g23.1, wu:fb74e02
Chromosome Location
Chr.15: 36941455 - 37027289 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099344.2 NP_001092814.2 13 - 14 2316 63
GenBank mRNA CU638818.1 - 13 - 14 1729 63
EF571006.1 - 13 - 14 2316 63

View Full Product Details