Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hint2
Entrez Gene ID
798688
Gene Name
histidine triad nucleotide binding protein 2
Gene Aliases
si:dkey-87i10.2, zgc:163082
Chromosome Location
Chr.5: 53009094 - 53067934 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083572.2 NP_001077041.1 1 - 2 127 68
GenBank mRNA BC134222.1 - 1 - 2 128 68
FJ032031.1 - 1 - 2 104 68

View Full Product Details