Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lingo4a
Entrez Gene ID
100005227
Gene Name
leucine rich repeat and Ig domain containing 4a
Gene Aliases
si:dkeyp-24a7.7
Chromosome Location
Chr.19: 294758 - 307715 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082978.1 NP_001076447.1 1 - 1 1554 75
XM_005159310.4 XP_005159367.1 3 - 3 2266 75

View Full Product Details