Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gys2
Entrez Gene ID
373082
Gene Name
glycogen synthase 2
Gene Aliases
cb765, zgc:112057
Chromosome Location
Chr.4: 16800022 - 16824324 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001018679.1 NP_001018199.1 2 - 3 508 57
GenBank mRNA BC095660.1 - 2 - 3 508 57

View Full Product Details