Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
anpepa
Entrez Gene ID
558452
Gene Name
alanyl (membrane) aminopeptidase a
Gene Aliases
fd60b03, si:ch211-200o3.4, wu:fd60b03
Chromosome Location
Chr.18: 58953 - 78975 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044861.1 NP_001038326.1 7 - 8 1418 74

View Full Product Details