Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tuba4l
Entrez Gene ID
327377
Gene Name
tubulin, alpha 4 like
Gene Aliases
fi05b01, wu:fi05b01, zgc:55727, zgc:76934
Chromosome Location
Chr.13: 6247252 - 6252513 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199795.2 NP_956089.1 3 - 4 517 90
GenBank mRNA AY398374.1 - 3 - 4 498 90
AY398431.1 - 3 - 4 517 90
BC046889.1 - 3 - 4 468 90
BC066482.1 - 3 - 4 496 90
BC164535.1 - 2 - 3 405 90

View Full Product Details