Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
heg1
Entrez Gene ID
406202
Gene Name
heart development protein with EGF-like domains 1
Gene Aliases
heg
Chromosome Location
Chr.9: 38684871 - 38707510 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212968.2 NP_998133.2 3 - 4 2040 70
GenBank mRNA AY507658.1 - 3 - 4 2041 70
AY507659.1 - 3 - 4 2041 70
AY507660.1 - 3 - 4 2041 70
BC162109.1 - 3 - 4 2039 70
BC162480.1 - 3 - 4 2039 70

View Full Product Details