Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cbwd
Entrez Gene ID
406536
Gene Name
COBW domain containing
Gene Aliases
fb76b04, fc73a04, wu:fb76b04, wu:fc73a04, zgc:77617
Chromosome Location
Chr.8: 29378 - 53722 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213253.1 NP_998418.1 11 - 12 831 112
GenBank mRNA BC065429.1 - 11 - 12 831 112

View Full Product Details