Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBXN7
Entrez Gene ID
424856
Gene Name
UBX domain protein 7
Gene Aliases
RNF168, UBXD7
Chromosome Location
Chr.9: 4891707 - 4916942 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015277123.2 XP_015132609.1 6 - 7 921 61
XM_015277124.2 XP_015132610.1 6 - 7 933 61
XM_422667.6 XP_422667.4 6 - 7 644 61
GenBank mRNA BX931318.2 - 6 - 7 633 61

View Full Product Details